4-Iodbenzaldehyd 97+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
392060010
392060050
ACRO392060010EA
136
EUR
ACRO392060010
ACRO392060050
4-Iodbenzaldehyd 97+%
4-Iodobenzaldehyde
Formel:
C₇H₅IO Schmelzpunkt: 77…78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00039576 CAS-Nummer: 15164-44-0 |
Spezifikation Testergebnisse
Appearance | White to yellow to beige Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 75 to 80 °C |
HPLC | ≥97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...