3-Hydroxyphenylacetylene 95+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
392410010
392410050
ACRO392410010EA
83.3
EUR
ACRO392410010
ACRO392410050
3-Hydroxyphenylacetylene 95+%
3-Hydroxyphenylacetylene
Formel:
C₈H₆O Dichte: 1,803 g/cm³ Flammpunkt: 103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078347 CAS-Nummer: 10401-11-3 |
Spezifikation Testergebnisse
Appearance | Clear yellow to orange to light brown Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.5840 to 1.5880 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...