di-Ammoniumhydrogencitrat 98%, rein

Lieferant: Thermo Fisher Scientific
Achtung

392505000 392500025 392501000
ACRO392505000EA 53 EUR
ACRO392505000 ACRO392500025 ACRO392501000
di-Ammoniumhydrogencitrat 98%, rein
di-Ammoniumhydrogencitrat
Formel: HOC(CO₂H)(CH₂CO₂NH₄)₂
Dichte: 1,5 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00013068
CAS-Nummer: 3012-65-5

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR