α-Brom-3-iodtoluol 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
α-Brom-m-iodtoluol, 3-Iodbenzylbromid
392530010
392530050
392530250
ACRO392530010EA
42.5
EUR
ACRO392530010
ACRO392530050
ACRO392530250
α-Brom-3-iodtoluol 97%
α-Brom-3-iodtoluol
Formel:
C₇H₆BrI Molecular Weight: 296.93 g/mol Schmelzpunkt: 47…50 °C Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019016 CAS-Nummer: 49617-83-6 UN: 3261 ADR: 8 II |
Spezifikation Testergebnisse
Appearance | Light orange to red to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 47 to 50 °C |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...