1-Bromisochinolin 95+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
392590050
392590010
ACRO392590050EA
329
EUR
ACRO392590050
ACRO392590010
1-Bromisochinolin 95+%
1-Bromoisoquinoline
Formel:
C₉H₆BrN Schmelzpunkt: 43…47 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234478 CAS-Nummer: 1532-71-4 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Yellow to brown Powder and/or chunks |
Infrared spectrum | Conforms |
Melting point | 42 to 48 °C |
HPLC | ≥95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...