Sand 40-100 mesh analysis_for

Lieferant: Thermo Fisher Scientific

Synonyme: Silicium(IV)oxid, Siliciumdioxid

393302500 393300010 393300050
ACRO393302500EA 36.4 EUR
ACRO393302500 ACRO393300010 ACRO393300050
Sand 40-100 mesh analysis_for
Sand
Formel: SiO₂
Molecular Weight: 60.08 g/mol
Siedepunkt: 2230 °C
Schmelzpunkt: 1610 °C
Dichte: 2.6
Lagertemperatur: Raumtemperatur
MDL: MFCD00011232
CAS-Nummer: 14808-60-7

Bestellen

Spezifikation Testergebnisse

Appearance Light beige to grey Crystals
Particle size (D50) 205 µm (Typical)
Aluminium (Al) ≤250 ppm (Typical)
Calcium (Ca) ≤80 ppm (Typical)
Chromium (Cr) ≤0.5 ppm (Typical)
Iron (Fe) ≤65 ppm (Typical)
Potassium (K) ≤50 ppm (Typical)
Titanium (Ti) ≤150 ppm (Typical)
Silicon dioxide (SiO2) ≥99.80 % (Typical)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR