(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 97%

Lieferant: Thermo Fisher Scientific

Synonyme: 1,2:5,6-Di-O-isopropylidene-D-mannitol, (1S,2S)-1,2-bis[(4R)-2,2-dimethyl-1,3-dioxolan-4-yl]ethane-1,2-diol, (1S,2S)-1,2-BIS((R)-2,2-DIMETHYL-1,3-DIOXOLAN-4-YL)ETHANE-1,2-DIOL

393330010
ACRO393330010EA 14.6 EUR
ACRO393330010
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol 97%
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol
Formel: C₁₂H₂₂O₆
Molecular Weight: 262.302 g/mol
Schmelzpunkt: 105…114 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00003211
CAS-Nummer: 1707-77-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Assay ≥96.0 %
Specific optical rotation +5° to +7° (20°C, 589 nm) c=5, chloroform


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR