(S)-1-(Pyridin-4-yl)ethanol 99+%, (99+% ee)

Lieferant: Thermo Fisher Scientific
Achtung

393370010 393372500
ACRO393370010EA 276 EUR
ACRO393370010 ACRO393372500
(S)-1-(Pyridin-4-yl)ethanol 99+%, (99+% ee)
(S)-1-(Pyridin-4-yl)ethanol

Appearance: White to cream Powder or crystals and/or chunks

Formel: C₇H₉NO
Schmelzpunkt: 67…70 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077866
CAS-Nummer: 54656-96-1

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 67 to 70 °C
GC ≥99.0 %
Enantiomeric excess ≥99.0 %
Specific optical rotation -56° to -61° (20 °C, 589 nm) (c=1, chloroform)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR