2,4,5-Trifluorphenylboronsäure 97%

Lieferant: Thermo Scientific
Achtung

393800050
ACRO393800050EA 128 EUR
ACRO393800050
2,4,5-Trifluorphenylboronsäure 97%
2,4,5-Trifluorphenylboronsäure
Formel: C₆H₄BF₃O₂
Molecular Weight: 175,9 g/mol
Schmelzpunkt: 118…119 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863165
CAS-Nummer: 247564-72-3

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR