4-Methoxy-2-methylphenylboronsäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methoxy-2-methylbenzeneboronic acid, (4-Methoxy-2-methylphenyl)boronic acid
393850010
393850050
ACRO393850010EA
56.9
EUR
ACRO393850010
ACRO393850050
4-Methoxy-2-methylphenylboronsäure 97%
4-Methoxy-2-methylphenylboronsäure
Formel:
C₈H₁₁BO₃ Molecular Weight: 165.99 g/mol Schmelzpunkt: 148…161 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02684315 CAS-Nummer: 208399-66-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...