N,N-Diethyl-1,4-phenylendiammoniumsulfat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N,N-Diethyl-p-phenylendiammoniumsulfat, N,N-Diethylbenzol-1,4-diammoniumsulfat

395000500 395000010 395002500
ACRO395000500EA 31.6 EUR
ACRO395000500 ACRO395000010 ACRO395002500
N,N-Diethyl-1,4-phenylendiammoniumsulfat 99%
N,N-Diethyl-1,4-phenylendiammoniumsulfat
Formel: C₁₀H₁₈N₂O₄S
Molecular Weight: 262.33 g/mol
Schmelzpunkt: 184…187 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012993
CAS-Nummer: 6283-63-2
EINECS: 228-500-6

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder or crystals and/or lumps
Infrared spectrum Conforms
Melting point 184 to 187 °C
Titration with NaOH ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR