Methyl-4-(aminomethyl)benzoathydrochlorid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
396780010
396780050
396780250
ACRO396780010EA
25.2
EUR
ACRO396780010
ACRO396780050
ACRO396780250
Methyl-4-(aminomethyl)benzoathydrochlorid 97%
Methyl-4-(aminomethyl)benzoathydrochlorid
Formel:
C₉H₁₁NO₂ Schmelzpunkt: 234,5…237 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00182671 CAS-Nummer: 6232-11-7 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals |
Infrared spectrum | Conforms |
Titration Argentometric | 96.0 to 104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...