(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
396790250
ACRO396790250EA
127
EUR
ACRO396790250
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 98%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride
Formel:
C₄H₉O₃N·HCl Schmelzpunkt: 152…161 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00066121 CAS-Nummer: 5874-57-7 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Specific optical rotation | -2.5° to -4.5° (20°C, 589 nm) (C=4, methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...