5-Aza-2'-deoxycytidine 98%

Lieferant: Thermo Fisher Scientific
Gefahr

396810100 396810500
ACRO396810100EA 162 EUR
ACRO396810100 ACRO396810500
5-Aza-2'-deoxycytidine 98%
5-Aza-2'-deoxycytidine

Appearance: White to off-white Powder

Formel: C₈H₁₂N₄O₄
Schmelzpunkt: 200 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00043011
CAS-Nummer: 2353-33-5
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR