2-Methyl-2-propanyl-(3-brompropyl)carbamat 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(tert-Butoxycarbonylamino)propyl bromide, 3-(Boc-amino)propyl bromide, tert-Butyl N-(3-Bromopropyl)carbamate, N-(3-Bromopropyl)carbamic acid tert-Butyl ester
397010010
397010050
ACRO397010010EA
151
EUR
ACRO397010010
ACRO397010050
2-Methyl-2-propanyl-(3-brompropyl)carbamat 95%
3-(Boc-amino)propylbromide
Appearance: White Low melting solid
Formel:
C8H16BrNO2 Schmelzpunkt: 34…36,5 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02683429 CAS-Nummer: 83948-53-2 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White Low melting solid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...