(4-Boc-aminophenyl)boronic acid 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (4-Boc-Aminophenyl)Boronic Acid, 4-Boc-aminophenylboronic acid, (4-Boc-Aminophenyl)boronic acid, 4-(N-BOC-amino)phenylboronic acid, [4-[(2-methylpropan-2-yl)oxycarbonylamino]phenyl]boronic Acid

397030010
ACRO397030010EA 69.2 EUR
ACRO397030010
(4-Boc-aminophenyl)boronic acid 97%
(4-Boc-aminophenyl)boronic acid
Formel: C₁₁H₁₆BNO₄
Molecular Weight: 237,062 g/mol
Schmelzpunkt: 199…204 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02093054
CAS-Nummer: 380430-49-9

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Crystalline powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR