di-Kaliumhydrogenphosphat 1.0 M in Wasser

Lieferant: Thermo Fisher Scientific

397080010 397080025
ACRO397080010EA 61.7 EUR
ACRO397080010 ACRO397080025
di-Kaliumhydrogenphosphat 1.0 M in Wasser
di-Kaliumhydrogenphosphat
Formel: K₂HPO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00011383
CAS-Nummer: 7758-11-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ee2dc83ce0381f3a1e1f9b57dbc5d1d8ba841eaa&vid=306136bcf5b49a54c1cce81ed487d64a353d0fd6&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Solution
Titration with HClO4 1.0 M Exact strength on the certificate of analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR