1H-Imidazol-4-carbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Imidazole-4-carboxylic acid

397110010 397110050
ACRO397110010EA 75.8 EUR
ACRO397110010 ACRO397110050
1H-Imidazol-4-carbonsäure 98%
1H-Imidazole-4-carboxylic acid
Formel: C₄H₄N₂O₂
Molecular Weight: 112.09 g/mol
Schmelzpunkt: 242…244 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082203
CAS-Nummer: 1072-84-0

Bestellen

Spezifikation Testergebnisse

Appearance Grey to tan Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR