Castor oil ethoxylated pH 6.0-8.1

Lieferant: Thermo Fisher Scientific

397282500 397280010
ACRO397282500EA 83.8 EUR
ACRO397282500 ACRO397280010
Castor oil ethoxylated pH 6.0-8.1
Castor oil ethoxylated
Siedepunkt: >100 °C (760 mmHg)
Schmelzpunkt: 4 °C
Dichte: 1,05 g/cm³
Flammpunkt: 257 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00130879
CAS-Nummer: 61791-12-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Viscous liquid
Infrared spectrum Conforms
Water ≤3.0 %
pH 6.0 to 8.0 (10 % in water)
Hydroxyl value 65 to 78 mg KOH/g
Iodine value 25 to 35 g I2/100 g
Saponification value 65 to 70 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR