4-Picolylbromid Hydrobromid 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-(Bromomethyl)pyridine hydrobromide

397330050 397330250
ACRO397330050EA 59 EUR
ACRO397330050 ACRO397330250
4-Picolylbromid Hydrobromid 97%
4-Picolylbromid Hydrobromid
Formel: C₆H₇Br₂N
Molecular Weight: 252.94 g/mol
Schmelzpunkt: 189…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01863545
CAS-Nummer: 73870-24-3
UN: 3261
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
Melting point 189 to 192 °C
Titration Argentometric ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR