1,3-Difluor-2-nitrobenzol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2,6-Difluoronitrobenzene
397340050
ACRO397340050EA
196
EUR
ACRO397340050
1,3-Difluor-2-nitrobenzol 98%
1,3-Difluor-2-nitrobenzol
Formel:
C₆H₃F₂NO₂ Molecular Weight: 159.09 g/mol Siedepunkt: 91…92 °C (11 mmHg) Dichte: 1.503 g/cm³ Flammpunkt: 88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00192035 CAS-Nummer: 19064-24-5 UN: 2810 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | Clear yellow to orange Liquid or low melting solid |
Infrared spectrum | Conforms |
GC | ≥97.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...