MES Natriumsalz 99%

Lieferant: Thermo Fisher Scientific

397350250 397351000 397355000
ACRO397350250EA 55.3 EUR
ACRO397350250 ACRO397351000 ACRO397355000
MES Natriumsalz 99%
MES Natriumsalz
Formel: C₆H₁₂NNaO₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD00065473
CAS-Nummer: 71119-23-8

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Infrared spectrum Conforms
Solubility (10 % in water) Clear colorless
UV at 280 nm A: ≤0.03
Titration after Ion exchange ≥98.5 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR