6-Bromindol 96%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
6-Bromo-1H-indole
397360010
397360050
397362500
ACRO397360010EA
130
EUR
ACRO397360010
ACRO397360050
ACRO397362500
6-Bromindol 96%
6-Bromindol
Formel:
C₈H₆BrN Schmelzpunkt: 92…96 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238550 CAS-Nummer: 52415-29-9 |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
Melting point | 92 to 96 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...