N-Acetylguanidine 97%

Lieferant: Thermo Fisher Scientific
Achtung

397440050 397440250
ACRO397440050EA 63.3 EUR
ACRO397440050 ACRO397440250
N-Acetylguanidine 97%
N-Acetylguanidine
Formel: C₃H₇N₃O
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075615
CAS-Nummer: 5699-40-1

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR