(S)-(+)-1-Boc-3-hydroxypyrrolidin 98+%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
tert-butyl (3S)-3-hydroxypyrrolidine-1-carboxylate, (S)-tert-Butyl 3-hydroxypyrrolidine-1-carboxylate
397580050
ACRO397580050EA
185
EUR
ACRO397580050
(S)-(+)-1-Boc-3-hydroxypyrrolidin 98+%
(S)-(+)-1-Boc-3-hydroxypyrrolidin
Formel:
C₉H₁₇NO₃ Molecular Weight: 187,2362 g/mol Schmelzpunkt: 60…64 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01317839 CAS-Nummer: 101469-92-5 |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder or low melting solid |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Specific optical rotation | +21° to +26° (20°C, 589 nm) (C=1, in Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...