3-Brom-4-fluorbenzoesäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
397700050
ACRO397700050EA
122
EUR
ACRO397700050
3-Brom-4-fluorbenzoesäure 98%
3-Bromo-4-fluorobenzoic acid
Formel:
C₇H₄BrFO₂ Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00042463 CAS-Nummer: 1007-16-5 |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 137 to 144 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...