Milrinon 97%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 6-Methyl-5-(4-pyridyl)-2-pyridone-3-carbonitrile, 2-Methyl-6-oxo-1,6-dihydro-[3,4'-bipyridine]-5-carbonitrile, 3-Cyano-6-methyl-5-(4-pyridyl)-2-pyridone

397870500
ACRO397870500EA 524 EUR
ACRO397870500
Milrinon 97%
Milrinon

Appearance: White to light pink Powder

Formel: C₁₂H₉N₃O
Molecular Weight: 211,22 g/mol
Schmelzpunkt: >280 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00133539
CAS-Nummer: 78415-72-2
UN: 3439
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light pink Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR