4-Brom-2-iodanilin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
397910050
ACRO397910050EA
154
EUR
ACRO397910050
4-Brom-2-iodanilin 97%
4-Brom-2-iodanilin
Formel:
C₆H₅BrIN Molecular Weight: 297,92 g/mol Schmelzpunkt: 69…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093764 CAS-Nummer: 66416-72-6 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Grey to purple Powder |
Infrared spectrum | Conforms |
Melting point | 68 to 72 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...