3-Cyan-4-fluorbenzaldehyd 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Fluoro-5-formylbenzonitrile
397940010
397940050
ACRO397940010EA
43.8
EUR
ACRO397940010
ACRO397940050
3-Cyan-4-fluorbenzaldehyd 99%
3-Cyano-4-fluorobenzaldehyde
Formel:
C₈H₄FNO Molecular Weight: 149.1219 g/mol Schmelzpunkt: 80…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863558 CAS-Nummer: 218301-22-5 |
Spezifikation Testergebnisse
Appearance | White to light orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 85 to 89 °C |
GC | ≥98.5 % |
NMR (1H-NMR) | Consistent with structure |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...