2-Nitroterephthalsäure-1-methylester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Methyl 2-nitroterephthalate

398000050
ACRO398000050EA 45 EUR
ACRO398000050
2-Nitroterephthalsäure-1-methylester 97%
2-Nitroterephthalsäure-1-methylester
Formel: C₉H₇NO₆
Molecular Weight: 225,17 g/mol
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024510
CAS-Nummer: 35092-89-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR