2-Chlor-4-hydroxybenzaldehyd 97%

Lieferant: Thermo Scientific
Achtung

398030010
ACRO398030010EA 138 EUR
ACRO398030010
2-Chlor-4-hydroxybenzaldehyd 97%
2-Chloro-4-hydroxybenzaldehyde
Formel: C₇H₅ClO₂
Molecular Weight: 156,57 g/mol
Schmelzpunkt: 145…148 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052184
CAS-Nummer: 56962-11-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to tan Powder
Infrared spectrum Conforms
Melting point 145 to 150 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR