6-Nitro-1,3-benzoxazol-2(3H)-one 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Nitrobenzoxazole-2(3H)-one
398150050
ACRO398150050EA
74.8
EUR
ACRO398150050
6-Nitro-1,3-benzoxazol-2(3H)-one 97%
6-Nitro-1,3-benzoxazol-2(3H)-one
Formel:
C₇H₄N₂O₄ Molecular Weight: 180,12 g/mol Schmelzpunkt: 244…247 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00463755 CAS-Nummer: 4694-91-1 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow to light grey Powder or solid |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...