2-Amino-m-kresol 96%

Lieferant: Thermo Fisher Scientific
Achtung

398410050 398410250
ACRO398410050EA 67.5 EUR
ACRO398410050 ACRO398410250
2-Amino-m-kresol 96%
2-Amino-m-kresol
Formel: C₇H₉NO
Schmelzpunkt: 149…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075082
CAS-Nummer: 2835-97-4

Bestellen

Spezifikation Testergebnisse

Appearance White to brown Crystalline powder or flakes
Infrared spectrum Conforms
Melting point 149 to 152 °C
HPLC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR