5-Brom-2-fluorbenzoesäure 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
398590050
ACRO398590050EA
77.9
EUR
ACRO398590050
5-Brom-2-fluorbenzoesäure 97%
5-Brom-2-fluorbenzoesäure
Formel:
C₇H₄BrFO₂ Schmelzpunkt: 141…145 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143423 CAS-Nummer: 146328-85-0 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 150 to 154 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...