(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: benzyl (2R,3S)-6-oxo-2,3-diphenylmorpholine-4-carboxylate, Benzyl (2R,3S)-(-)-6-oxo-2,3-diphenyl-4-morpholinecarboxylate, (2R,3S)-Benzyl 6-oxo-2,3-diphenylmorpholine-4-carboxylate

398660010
ACRO398660010EA 96.9 EUR
ACRO398660010
(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone 98%
(5R,6S)-(-)-4-Benzyloxycarbonyl-5,6-diphenyl-2-morpholinone
Formel: C₂₄H₂₁NO₄
Molecular Weight: 387,435 g/mol
Schmelzpunkt: 205…207 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00074958
CAS-Nummer: 100516-54-9

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -64° to -68° (25°C, 589 nm) (c=5.5,CH2Cl2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR