ω-Brom-2-nitroacetophenon 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Brommethyl-o-nitrophenylketon, 2-Brom-2'-nitroacetophenon, α-Brom-2-nitroacetophenon, 2-Nitrophenacylbromid
398850050
ACRO398850050EA
124
EUR
ACRO398850050
ω-Brom-2-nitroacetophenon 98%
ω-Brom-2-nitroacetophenon
Formel:
C₈H₆BrNO₃ Molecular Weight: 244,04 g/mol Schmelzpunkt: 55…57 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010294 CAS-Nummer: 6851-99-6 UN: 3261 ADR: 8,II |
Spezifikation Testergebnisse
Appearance | White to light yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 53 to 58 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...