(4S)-4-Hydroxy-2-pyrrolidinon 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(4S)-(-)-beta-Hydroxy-gamma-butyrolactam, (S)-4-Hydroxy-2-pyrrolidone, (S)-(-)-4-Hydroxy-2-pyrrolidinone
398970050
ACRO398970050EA
235
EUR
ACRO398970050
(4S)-4-Hydroxy-2-pyrrolidinon 98%
(S)-4-Hydroxypyrrolidine-2-one
Formel:
C₄H₇NO₂ Molecular Weight: 101,105 g/mol Schmelzpunkt: 156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273370 CAS-Nummer: 68108-18-9 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 156 to 159 °C |
GC | ≥97.5 % |
Specific optical rotation | -40° to -46° (20 °C, 589 nm) (c=1, EtOH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...