Ethyltrifluorpyruvat 98+%

Lieferant: Thermo Scientific
Achtung

Synonyme: Ethyl 3,3,3-trifluoro-2-oxopropanoate, Ethyl 3,3,3-trifluoropyruvate

399010050
ACRO399010050EA 53.1 EUR
ACRO399010050
Ethyltrifluorpyruvat 98+%
Ethyltrifluorpyruvat
Formel: C₅H₅F₃O₃
Molecular Weight: 170,09 g/mol
Flammpunkt: 27 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00114935
CAS-Nummer: 13081-18-0
UN: 3272
ADR: 3,III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3390 to 1.3410 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR