1,4-Dihydro-2,6-dimethylpyridin-3,5-dicarbonsäure-diethylester 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Hantzsch Ester, 1,4-Dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester, Diethyl 1,4-dihydro-3,5-pyridinecarboxylate, Etidin, Diethyl 1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
399190010
ACRO399190010EA
73.8
EUR
ACRO399190010
1,4-Dihydro-2,6-dimethylpyridin-3,5-dicarbonsäure-diethylester 95%
Diethyl-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate
Formel:
C₁₃H₁₉NO₄ Molecular Weight: 253,30 g/mol Schmelzpunkt: 178…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005951 CAS-Nummer: 1149-23-1 |
Spezifikation Testergebnisse
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...