4-(2-Pyridyl)benzaldehyd 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Pyridin-2-yl-benzaldehyde, 4-(Pyridin-2-yl)benzaldehyde
399240050
ACRO399240050EA
113
EUR
ACRO399240050
4-(2-Pyridyl)benzaldehyd 97%
4-(2-Pyridyl)benzaldehyd
Formel:
C₁₂H₉NO Molecular Weight: 183,21 g/mol Schmelzpunkt: 51…53 °C Flammpunkt: 113 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863537 CAS-Nummer: 127406-56-8 |
Spezifikation Testergebnisse
Appearance | White to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 48 to 54 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...