N-Boc-D-serine methyl ester 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: N-(tert-Butoxycarbonyl)-D-serine methyll ester, Boc-D-Ser-OMe, N-Boc-D-serine methyll ester

399370100
ACRO399370100EA 156 EUR
ACRO399370100
N-Boc-D-serine methyl ester 97%
N-Boc-D-serine methyl ester

Appearance: Clear colorless to yellow Viscous liquid

Formel: C₉H₁₇NO₅
Molecular Weight: 219,237 g/mol
Siedepunkt: 215 °C (760 mmHg)
Dichte: 1,08 g/cm³
Flammpunkt: 113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00270516
CAS-Nummer: 95715-85-8

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
GC ≥96.0 %
Specific optical rotation +16° to +20° (20°C, 589 nm) (c=5% in Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR