6-Chloroxindol 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Chloro-2-oxindole

399440250
ACRO399440250EA 120 EUR
ACRO399440250
6-Chloroxindol 98%
6-Chloroxindol
Formel: C₈H₆ClNO
Molecular Weight: 167,59 g/mol
Schmelzpunkt: 195…199 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00209962
CAS-Nummer: 56341-37-8

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to tan Crystalline powder
Infrared spectrum Conforms
Melting point 195 to 199 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR