tert-Butyl-1,4-diazepane-1-carboxylate 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1-BOC-hexahydro-1,4-diazepine, tert-Butyl 1,4-diazepane-1-carboxylate

399530050
ACRO399530050EA 73.7 EUR
ACRO399530050
tert-Butyl-1,4-diazepane-1-carboxylate 98%
tert-Butyl-1,4-diazepane-1-carboxylate

Appearance: Colorless to yellow Liquid

Formel: C₁₀H₂₀N₂O₂
Molecular Weight: 200,28 g/mol
Siedepunkt: 92…96 °C (,5 mmHg)
Dichte: 1,029 g/cm³
Flammpunkt: >110 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276987
CAS-Nummer: 112275-50-0
UN: 2735
ADR: 8,III

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR