1-Boc-piperidin-3-carbonsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid

399580050 399580250
ACRO399580050EA 36 EUR
ACRO399580050 ACRO399580250
1-Boc-piperidin-3-carbonsäure 98%
1-Boc-piperidin-3-carbonsäure
Formel: C₁₁H₁₉NO₄
Schmelzpunkt: 159…160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673775
CAS-Nummer: 84358-12-3

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 159 to 162 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR