1-Boc-piperidin-3-carbonsäure 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
N-Boc-DL-nipecotic acid, 1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid, 1-Boc-Nipecotic acid, Boc-Nip-OH, 1-(tert-Butoxycarbonyl)piperidine-3-carboxylic acid, N-BOC-3-Piperidinecarboxylic acid
399580050
399580250
ACRO399580050EA
36
EUR
ACRO399580050
ACRO399580250
1-Boc-piperidin-3-carbonsäure 98%
1-Boc-piperidin-3-carbonsäure
Formel:
C₁₁H₁₉NO₄ Schmelzpunkt: 159…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00673775 CAS-Nummer: 84358-12-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 159 to 162 °C |
Titration with NaOH | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...