1-Acetylindole-3-carboxaldehyde 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Acetyl-3-indolecarboxaldehyde, 1-Acetyl-1H-indole-3-carbaldehyde

399700050
ACRO399700050EA 67.1 EUR
ACRO399700050
1-Acetylindole-3-carboxaldehyde 98%
1-Acetylindole-3-carboxaldehyde
Formel: C₁₁H₉NO₂
Molecular Weight: 187,20 g/mol
Schmelzpunkt: 165 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039691
CAS-Nummer: 22948-94-3

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 163 to 167 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR