1-Chloroisoquinolin-4-ol 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Chloro-4-hydroxyisoquinoline

399780010
ACRO399780010EA 111 EUR
ACRO399780010
1-Chloroisoquinolin-4-ol 97%
1-Chloroisoquinolin-4-ol
Formel: C₉H₆ClNO
Molecular Weight: 179,61 g/mol
Schmelzpunkt: 195…197 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425941
CAS-Nummer: 3336-43-4
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to light pink Crystalline powder
Infrared spectrum Conforms
Melting point 184 to 192 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR