5-Brom-1H-indol-3-carbaldehyd 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Bromoindole-3-carboxaldehyde, 5-Bromo-3-formylindole

399790050
ACRO399790050EA 73.2 EUR
ACRO399790050
5-Brom-1H-indol-3-carbaldehyd 97%
5-Bromo-1H-indole-3-carbaldehyde

Appearance: White to pink to light brown Powder

Formel: C₉H₆BrNO
Molecular Weight: 224,06 g/mol
Schmelzpunkt: 205…206 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00152016
CAS-Nummer: 877-03-2

Bestellen

Spezifikation Testergebnisse

Appearance White to pink to light brown Powder
Infrared spectrum Conforms
Melting point 200 to 207 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR