3-Brom-1-(trimethylsilyl)-1-propin 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(Trimethylsilyl)propargyl bromide, (3-Bromoprop-1-yn-1-yl)trimethylsilane
399840010
399840050
ACRO399840010EA
41.1
EUR
ACRO399840010
ACRO399840050
3-Brom-1-(trimethylsilyl)-1-propin 97%
3-Brom-1-(trimethylsilyl)-1-propin
Formel:
C₆H₁₁BrSi Molecular Weight: 191.14 g/mol Siedepunkt: 44…45 °C (2 mmHg) Dichte: 1.351 g/cm³ Flammpunkt: 62 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00134460 CAS-Nummer: 38002-45-8 |
Spezifikation Testergebnisse
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4820 to 1.4840 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...