6-Methoxy-2-naphthalinboronsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid

399860050
ACRO399860050EA 76.7 EUR
ACRO399860050
6-Methoxy-2-naphthalinboronsäure 97%
6-Methoxy-2-naphthaleneboronic acid
Formel: C₁₁H₁₁BO₃
Molecular Weight: 202,02 g/mol
Schmelzpunkt: >295 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03093087
CAS-Nummer: 156641-98-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white to light brown Powder or crystalline powder
Infrared spectrum Authentic
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR